2'-Bromo-6'-fluoro-3'-methoxy-[1,1'-biphenyl]-4-carbaldehyde ≥97%

Supplier: AOBChem USA

Warning

68204-500MG 68204-1G 68204-5G 68204-25G
77333-170EA 121.54 USD
77333-170 77343-874 77369-198 77381-288
2'-Bromo-6'-fluoro-3'-methoxy-[1,1'-biphenyl]-4-carbaldehyde ≥97%
2'-Bromo-6'-fluoro-3'-methoxy-[1,1'-biphenyl]-4-carbaldehyde

Precaución: for Research Use Only

Formula: C₁₄H₁₀BrFO₂
MW: 309.13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34763236
CAS Number: 2901102-05-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR