2-Bromo-6-fluoro-3,4'-dimethoxy-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

68203-500MG 68203-1G 68203-5G 68203-25G
77333-168EA 128.75 USD
77333-168 77344-284 77369-196 77381-286
2-Bromo-6-fluoro-3,4'-dimethoxy-1,1'-biphenyl ≥97%
2-Bromo-6-fluoro-3,4'-dimethoxy-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₄H₁₂BrFO₂
Storage Temperature: Ambient
MDL Number: MFCD34763235
CAS Number: 2901083-21-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR