2-Bromo-4',6-difluoro-3-methoxy-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

68202-500MG 68202-1G 68202-5G 68202-25G
77333-166EA 103 USD
77333-166 77343-048 77369-194 77381-284
2-Bromo-4',6-difluoro-3-methoxy-1,1'-biphenyl ≥97%
2-Bromo-4',6-difluoro-3-methoxy-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₃H₉BrF₂O
MW: 299.111 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34763234
CAS Number: 2901099-08-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR