2-Bromo-6-fluoro-3-methoxy-4'-methyl-1,1'-biphenyl ≥97%
Supplier: AOBChem USA
|
Warning
|
68198-500MG
68198-1G
68198-5G
68198-25G
77333-162EA
101.97
USD
77333-162
77343-012
77369-190
77381-280
2-Bromo-6-fluoro-3-methoxy-4'-methyl-1,1'-biphenyl ≥97%
2-Bromo-6-fluoro-3-methoxy-4'-methyl-1,1'-biphenyl
Precaución: for Research Use Only
Formula:
C₁₄H₁₂BrFO Storage Temperature: Ambient |
MDL Number:
MFCD34763231 CAS Number: 2901081-52-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...