2'-Methoxy-5'-(trifluoromethyl)-[1,1'-biphenyl]-4-carboxylic acid ≥97%

Supplier: AOBChem USA

Warning

68176-500MG 68176-1G 68176-5G
77340-046EA 723.06 USD
77340-046 77357-362 77381-274
2'-Methoxy-5'-(trifluoromethyl)-[1,1'-biphenyl]-4-carboxylic acid ≥97%
2'-Methoxy-5'-(trifluoromethyl)-[1,1'-biphenyl]-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₅H₁₁F₃O₃
MW: 296.241 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18312746
CAS Number: 1261998-93-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR