1-(Benzyloxy)-2-naphthaldehyde ≥97%

Supplier: AOBChem USA

Warning

68171-500MG 68171-1G 68171-5G 68171-25G
77333-158EA 152.44 USD
77333-158 77345-492 77369-186 77390-270
1-(Benzyloxy)-2-naphthaldehyde ≥97%
1-(Benzyloxy)-2-naphthaldehyde

Precaución: for Research Use Only

Formula: C₁₈H₁₄O₂
Storage Temperature: Ambient
MDL Number: MFCD34763209
CAS Number: 109380-65-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR