1-(Benzyloxy)-2-bromonaphthalene ≥97%

Supplier: AOBChem USA

Warning

68167-1G 68167-5G 68167-25G
77340-808EA 60.77 USD
77340-808 77369-182 77381-270
1-(Benzyloxy)-2-bromonaphthalene ≥97%
1-(Benzyloxy)-2-bromonaphthalene

Precaución: for Research Use Only

Formula: C₁₇H₁₃BrO
MW: 313.189 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34763206
CAS Number: 76939-81-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR