1-Isopropoxy-2-naphthaldehyde ≥97%

Supplier: AOBChem USA

Warning

68165-500MG 68165-1G 68165-25G 68165-5G
77333-154EA 297.67 USD
77333-154 77357-358 77390-266 77381-268
1-Isopropoxy-2-naphthaldehyde ≥97%
1-Isopropoxy-2-naphthaldehyde

Precaución: for Research Use Only

Formula: C₁₄H₁₄O₂
MW: 214.26 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34763204
CAS Number: 1093352-68-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR