1-Isopropoxy-2-naphthoic acid ≥97%

Supplier: AOBChem USA

Warning

68163-500MG 68163-1G 68163-5G 68163-25G
77333-152EA 281.19 USD
77333-152 77357-356 77369-180 77390-264
1-Isopropoxy-2-naphthoic acid ≥97%
1-Isopropoxy-2-naphthoic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₄O₃
MW: 230.259 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13475927
CAS Number: 172088-71-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR