2-Bromo-1-isopropoxynaphthalene ≥97%

Supplier: AOBChem USA

Warning

68160-500MG 68160-1G 68160-5G 68160-25G
77333-148EA 95.79 USD
77333-148 77342-626 77369-176 77381-266
2-Bromo-1-isopropoxynaphthalene ≥97%
2-Bromo-1-isopropoxynaphthalene

Precaución: for Research Use Only

Formula: C₁₃H₁₃BrO
MW: 265.146 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34763200
CAS Number: 2807455-09-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR