1-(5-Bromo-2,3-difluoro-4-methylphenyl)-2,2,2-trifluoroethanone ≥97%

Supplier: AOBChem USA

Warning

68156-500MG 68156-1G 68156-5G 68156-25G
77333-146EA 141.11 USD
77333-146 77344-890 77369-174 77390-260
1-(5-Bromo-2,3-difluoro-4-methylphenyl)-2,2,2-trifluoroethanone ≥97%
1-(5-Bromo-2,3-difluoro-4-methylphenyl)-2,2,2-trifluoroethanone

Precaución: for Research Use Only

Formula: C₉H₄BrF₅O
MW: 303.023 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34763199
CAS Number: 2807464-48-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR