5-(3,4,5-Trimethoxyphenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

68125-500MG 68125-1G 68125-5G 68125-25G
77333-136EA 161.71 USD
77333-136 77345-964 77369-166 77390-252
5-(3,4,5-Trimethoxyphenyl)thiazol-2-amine ≥97%
5-(3,4,5-Trimethoxyphenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₂H₁₄N₂O₃S
MW: 266.316 g/mol
Storage Temperature: Ambient
CAS Number: 1681084-05-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR