4-Bromo-1-methoxy-2-propoxybenzene ≥95%
Supplier: AOBChem USA
|
Warning
|
68121-500MG
68121-1G
68121-5G
68121-25G
77333-134EA
163.77
USD
77333-134
77346-124
77369-164
77390-250
4-Bromo-1-methoxy-2-propoxybenzene ≥95%
4-Bromo-1-methoxy-2-propoxybenzene
Precaución: for Research Use Only
Formula:
C₁₀H₁₃BrO₂ MW: 245.113 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD27952919 CAS Number: 1823919-71-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...