5-(4-(Pentyloxy)phenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

68116-500MG 68116-1G 68116-5G 68116-25G
77333-132EA 141.11 USD
77333-132 77344-886 77369-162 77390-248
5-(4-(Pentyloxy)phenyl)thiazol-2-amine ≥97%
5-(4-(Pentyloxy)phenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₄H₁₈N₂OS
MW: 262.371 g/mol
Storage Temperature: Ambient
CAS Number: 2901085-08-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR