(3,5-Dibromo-2-fluorophenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

68110-500MG 68110-1G 68110-5G 68110-25G
77333-130EA 281.19 USD
77333-130 77357-346 77369-160 77390-246
(3,5-Dibromo-2-fluorophenyl)methanol ≥97%
(3,5-Dibromo-2-fluorophenyl)methanol

Precaución: for Research Use Only

Formula: C₇H₅Br₂FO
MW: 283.92 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03094097
CAS Number: 497181-27-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR