1-Chloro-3-ethyl-2-fluorobenzene ≥97%

Supplier: AOBChem USA

Warning

68097-500MG 68097-1G 68097-5G
77339-796EA 643.75 USD
77339-796 77357-344 77381-260
1-Chloro-3-ethyl-2-fluorobenzene ≥97%
1-Chloro-3-ethyl-2-fluorobenzene

Precaución: for Research Use Only

Formula: C₈H₈ClF
Storage Temperature: Ambient
MDL Number: MFCD22490980
CAS Number: 1369790-27-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR