3-Iodo-4-methyl-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

68068-500MG 68068-1G 68068-5G 68068-25G
77333-124EA 204.97 USD
77333-124 77357-338 77369-154 77390-240
3-Iodo-4-methyl-1,1'-biphenyl ≥97%
3-Iodo-4-methyl-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₃H₁₁I
Storage Temperature: Ambient
MDL Number: MFCD34763170
CAS Number: 1890112-82-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR