(3'-Bromo-5'-(trifluoromethoxy)-[1,1'-biphenyl]-4-yl)methanol ≥95%

Supplier: AOBChem USA

Warning

68053-1G 68053-5G 68053-25G
77340-962EA 64.89 USD
77340-962 77369-152 77381-248
(3'-Bromo-5'-(trifluoromethoxy)-[1,1'-biphenyl]-4-yl)methanol ≥95%
(3'-Bromo-5'-(trifluoromethoxy)-[1,1'-biphenyl]-4-yl)methanol

Precaución: for Research Use Only

Formula: C₁₄H₁₀BrF₃O₂
Storage Temperature: Ambient
MDL Number: MFCD34763155
CAS Number: 2901106-76-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR