3'-Bromo-5'-(trifluoromethoxy)-[1,1'-biphenyl]-4-carbaldehyde ≥97%

Supplier: AOBChem USA

Warning

68052-1G 68052-5G 68052-25G
77341-252EA 78.28 USD
77341-252 77369-150 77381-246
3'-Bromo-5'-(trifluoromethoxy)-[1,1'-biphenyl]-4-carbaldehyde ≥97%
3'-Bromo-5'-(trifluoromethoxy)-[1,1'-biphenyl]-4-carbaldehyde

Precaución: for Research Use Only

Formula: C₁₄H₈BrF₃O₂
MW: 345.111 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34763154
CAS Number: 2901067-03-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR