3-Bromo-4',5-bis(trifluoromethoxy)-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

68049-1G 68049-5G 68049-25G
77341-888EA 106.09 USD
77341-888 77369-148 77381-244
3-Bromo-4',5-bis(trifluoromethoxy)-1,1'-biphenyl ≥97%
3-Bromo-4',5-bis(trifluoromethoxy)-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₄H₇BrF₆O₂
Storage Temperature: Ambient
MDL Number: MFCD34763151
CAS Number: 2901066-94-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR