3-Bromo-3'-methoxy-5-(trifluoromethoxy)-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

68044-1G 68044-5G 68044-25G
77341-604EA 94.76 USD
77341-604 77369-144 77381-240
3-Bromo-3'-methoxy-5-(trifluoromethoxy)-1,1'-biphenyl ≥97%
3-Bromo-3'-methoxy-5-(trifluoromethoxy)-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₄H₁₀BrF₃O₂
MW: 347.127 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34763146
CAS Number: 2901102-38-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR