3-Bromo-4'-methoxy-5-(trifluoromethoxy)-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

68038-1G 68038-5G 68038-25G
77341-720EA 101.97 USD
77341-720 77369-140 77381-236
3-Bromo-4'-methoxy-5-(trifluoromethoxy)-1,1'-biphenyl ≥97%
3-Bromo-4'-methoxy-5-(trifluoromethoxy)-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₄H₁₀BrF₃O₂
MW: 347.127 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34763140
CAS Number: 2901097-27-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR