3-Bromo-4'-fluoro-5-(trifluoromethoxy)-1,1'-biphenyl ≥95%

Supplier: AOBChem USA

Warning

68035-500MG 68035-1G 68035-5G 68035-25G
77333-118EA 116.39 USD
77333-118 77343-504 77369-138 77381-234
3-Bromo-4'-fluoro-5-(trifluoromethoxy)-1,1'-biphenyl ≥95%
3-Bromo-4'-fluoro-5-(trifluoromethoxy)-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₃H₇BrF₄O
MW: 335.092 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34763137
CAS Number: 2901098-66-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR