3-Bromo-4'-methyl-5-(trifluoromethoxy)-1,1'-biphenyl ≥95%

Supplier: AOBChem USA

Warning

68032-1G 68032-5G 68032-25G
77341-250EA 78.28 USD
77341-250 77369-136 77381-232
3-Bromo-4'-methyl-5-(trifluoromethoxy)-1,1'-biphenyl ≥95%
3-Bromo-4'-methyl-5-(trifluoromethoxy)-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₄H₁₀BrF₃O
MW: 331.128 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34763134
CAS Number: 2901085-03-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR