3'-Bromo-2-methyl-5'-(trifluoromethoxy)-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

68026-1G 68026-5G 68026-25G
77341-234EA 77.25 USD
77341-234 77369-132 77381-228
3'-Bromo-2-methyl-5'-(trifluoromethoxy)-1,1'-biphenyl ≥97%
3'-Bromo-2-methyl-5'-(trifluoromethoxy)-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₄H₁₀BrF₃O
MW: 331.128 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34763128
CAS Number: 2901102-06-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR