3-Chloro-2-(methylthio)-4-propoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

68022-500MG 68022-1G 68022-5G
77335-946EA 369.77 USD
77335-946 77357-326 77381-224
3-Chloro-2-(methylthio)-4-propoxybenzoic acid ≥97%
3-Chloro-2-(methylthio)-4-propoxybenzoic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₃ClO₃S
MW: 260.737 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34763124
CAS Number: 2901098-95-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR