Methyl 5-fluoro-4-isopropoxy-2-(trifluoromethyl)benzoate ≥97%

Supplier: AOBChem USA

Warning

68017-500MG 68017-1G 68017-5G
77333-112EA 311.06 USD
77333-112 77357-322 77381-222
Methyl 5-fluoro-4-isopropoxy-2-(trifluoromethyl)benzoate ≥97%
Methyl 5-fluoro-4-isopropoxy-2-(trifluoromethyl)benzoate

Precaución: for Research Use Only

Formula: C₁₂H₁₂F₄O₃
MW: 280.215 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34763119
CAS Number: 2918889-00-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR