4-Bromo-3-fluoro-5-methoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

68000-500MG 68000-1G 68000-5G 68000-25G
77333-108EA 276.04 USD
77333-108 77357-308 77369-126 77390-234
4-Bromo-3-fluoro-5-methoxybenzaldehyde ≥97%
4-Bromo-3-fluoro-5-methoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₆BrFO₂
MW: 233.034 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34763102
CAS Number: 1783692-34-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR