2-Fluoro-4-formyl-6-methoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

67995-500MG 67995-1G 67995-5G
77333-104EA 336.81 USD
77333-104 77357-306 77381-210
2-Fluoro-4-formyl-6-methoxybenzoic acid ≥97%
2-Fluoro-4-formyl-6-methoxybenzoic acid

Precaución: for Research Use Only

Formula: C₉H₇FO₄
MW: 198.148 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34763100
CAS Number: 2386964-17-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR