1-Bromo-3-chloro-4-methoxy-2-(methylsulfonyl)benzene ≥97%

Supplier: AOBChem USA

Warning

67994-500MG 67994-1G 67994-5G 67994-25G
77333-102EA 157.59 USD
77333-102 77345-716 77369-122 77390-232
1-Bromo-3-chloro-4-methoxy-2-(methylsulfonyl)benzene ≥97%
1-Bromo-3-chloro-4-methoxy-2-(methylsulfonyl)benzene

Precaución: for Research Use Only

Formula: C₈H₈BrClO₃S
MW: 299.569 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34763099
CAS Number: 2807448-55-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR