2-(Difluoromethyl)-4-methoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

67984-500MG 67984-1G 67984-5G
77336-274EA 384.19 USD
77336-274 77357-296 77381-206
2-(Difluoromethyl)-4-methoxybenzoic acid ≥97%
2-(Difluoromethyl)-4-methoxybenzoic acid

Precaución: for Research Use Only

Formula: C₉H₈F₂O₃
MW: 202.155 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32065891
CAS Number: 2248315-12-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR