4'-Methyl-4-nitro-3-(trifluoromethyl)-1,1'-biphenyl ≥95%
Supplier: AOBChem USA
|
Warning
|
67977-500MG
67977-1G
67977-5G
67977-25G
77333-092EA
182.31
USD
77333-092
77347-012
77369-114
77390-224
4'-Methyl-4-nitro-3-(trifluoromethyl)-1,1'-biphenyl ≥95%
4'-Methyl-4-nitro-3-(trifluoromethyl)-1,1'-biphenyl
Precaución: for Research Use Only
Formula:
C₁₄H₁₀F₃NO₂ MW: 281.23 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD34763091 CAS Number: 2055940-54-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...