4-Phenylbenzo[d][1,3]dioxole-5-carboxylic acid ≥97%

Supplier: AOBChem USA

Warning

67967-500MG 67967-1G 67967-5G
77338-228EA 469.68 USD
77338-228 77357-290 77381-198
4-Phenylbenzo[d][1,3]dioxole-5-carboxylic acid ≥97%
4-Phenylbenzo[d][1,3]dioxole-5-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₀O₄
MW: 242.227 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34763084
CAS Number: 959058-63-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR