5-Bromo-4-phenylbenzo[d][1,3]dioxole ≥95%

Supplier: AOBChem USA

Warning

67964-500MG 67964-1G 67964-5G 67964-25G
77333-082EA 178.19 USD
77333-082 77346-686 77369-106 77390-216
5-Bromo-4-phenylbenzo[d][1,3]dioxole ≥95%
5-Bromo-4-phenylbenzo[d][1,3]dioxole

Precaución: for Research Use Only

Formula: C₁₃H₉BrO₂
MW: 277.113 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34763083
CAS Number: 2807444-70-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR