4-Bromo-2-chloro-3-phenylpyridine ≥95%

Supplier: AOBChem USA

Warning

67931-500MG 67931-1G 67931-5G
77337-216EA 408.91 USD
77337-216 77357-282 77381-192
4-Bromo-2-chloro-3-phenylpyridine ≥95%
4-Bromo-2-chloro-3-phenylpyridine

Precaución: for Research Use Only

Formula: C₁₁H₇BrClN
Storage Temperature: Ambient
MDL Number: MFCD34763072
CAS Number: 2807441-12-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR