4-Bromo-3,4'-dimethoxy-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

67929-500MG 67929-1G 67929-5G 67929-25G
77333-078EA 128.75 USD
77333-078 77344-282 77369-102 77381-190
4-Bromo-3,4'-dimethoxy-1,1'-biphenyl ≥97%
4-Bromo-3,4'-dimethoxy-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₄H₁₃BrO₂
Storage Temperature: Ambient
MDL Number: MFCD34763071
CAS Number: 2807449-74-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR