4-Bromo-3-methoxy-3'-methyl-1,1'-biphenyl ≥95%

Supplier: AOBChem USA

Warning

67927-500MG 67927-1G 67927-5G 67927-25G
77333-074EA 184.37 USD
77333-074 77347-314 77369-098 77390-210
4-Bromo-3-methoxy-3'-methyl-1,1'-biphenyl ≥95%
4-Bromo-3-methoxy-3'-methyl-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₄H₁₃BrO
MW: 277.156 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34763069
CAS Number: 2807441-90-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR