4'-Bromo-3'-methoxy-2,3-dimethyl-1,1'-biphenyl ≥95%

Supplier: AOBChem USA

Warning

67926-500MG 67926-1G 67926-5G 67926-25G
77333-072EA 161.71 USD
77333-072 77345-962 77369-096 77390-208
4'-Bromo-3'-methoxy-2,3-dimethyl-1,1'-biphenyl ≥95%
4'-Bromo-3'-methoxy-2,3-dimethyl-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₅H₁₅BrO
Storage Temperature: Ambient
MDL Number: MFCD34763068
CAS Number: 2807449-84-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR