4'-Bromo-3'-methoxy-2-methyl-1,1'-biphenyl ≥95%

Supplier: AOBChem USA

Warning

67925-1G 67925-5G 67925-25G
77340-806EA 60.77 USD
77340-806 77369-094 77381-188
4'-Bromo-3'-methoxy-2-methyl-1,1'-biphenyl ≥95%
4'-Bromo-3'-methoxy-2-methyl-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₄H₁₃BrO
MW: 277.156 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34763067
CAS Number: 2807448-27-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR