3-Methoxy-4'-methyl-[1,1'-biphenyl]-4-carboxylic acid ≥97%

Supplier: AOBChem USA

Warning

67918-500MG 67918-1G 67918-5G 67918-25G
77333-068EA 202.91 USD
77333-068 77357-278 77369-092 77390-204
3-Methoxy-4'-methyl-[1,1'-biphenyl]-4-carboxylic acid ≥97%
3-Methoxy-4'-methyl-[1,1'-biphenyl]-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₅H₁₄O₃
MW: 242.27 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18318964
CAS Number: 175153-28-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR