3-Fluoro-2-isopropoxy-5-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

67912-500MG 67912-1G 67912-5G
77337-814EA 445.99 USD
77337-814 77357-274 77381-184
3-Fluoro-2-isopropoxy-5-methylbenzaldehyde ≥97%
3-Fluoro-2-isopropoxy-5-methylbenzaldehyde

Precaución: for Research Use Only

Formula: C₁₁H₁₃FO₂
MW: 196.218 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28626732
CAS Number: 1557942-85-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR