3-Fluoro-2-isopropoxy-5-methylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

67911-500MG 67911-1G 67911-5G 67911-25G
77333-064EA 252.35 USD
77333-064 77357-272 77369-088 77390-200
3-Fluoro-2-isopropoxy-5-methylbenzoic acid ≥97%
3-Fluoro-2-isopropoxy-5-methylbenzoic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₃FO₃
MW: 212.218 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28627392
CAS Number: 1550665-55-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR