2,2-Difluoro-1-(naphthalen-1-yl)ethanone ≥97%

Supplier: AOBChem USA

Warning

67909-500MG 67909-1G 67909-5G 67909-25G
77333-060EA 210.12 USD
77333-060 77357-268 77369-084 77390-196
2,2-Difluoro-1-(naphthalen-1-yl)ethanone ≥97%
2,2-Difluoro-1-(naphthalen-1-yl)ethanone

Precaución: for Research Use Only

Formula: C₁₂H₈F₂O
MW: 206.188 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13173095
CAS Number: 715-80-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR