2,2-Difluoro-1-(4-(trifluoromethyl)phenyl)ethanone ≥95%
Supplier: AOBChem USA
|
Warning
|
67903-500MG
67903-1G
67903-5G
67903-25G
77333-058EA
231.75
USD
77333-058
77357-266
77369-082
77390-194
2,2-Difluoro-1-(4-(trifluoromethyl)phenyl)ethanone ≥95%
2,2-Difluoro-1-(4-(trifluoromethyl)phenyl)ethanone
Precaución: for Research Use Only
Formula:
C₉H₅F₅O Storage Temperature: Ambient |
MDL Number:
MFCD16618059 CAS Number: 1198355-60-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...