1-(2-Chloro-3-fluorophenyl)-2,2-difluoroethanone ≥95%

Supplier: AOBChem USA

Warning

67900-500MG 67900-1G 67900-5G
77336-936EA 400.67 USD
77336-936 77357-264 77381-182
1-(2-Chloro-3-fluorophenyl)-2,2-difluoroethanone ≥95%
1-(2-Chloro-3-fluorophenyl)-2,2-difluoroethanone

Precaución: for Research Use Only

Formula: C₈H₄ClF₃O
MW: 208.565 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28105479
CAS Number: 1822674-37-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR