1-(2,5-Difluoro-4-methoxyphenyl)-2,2-difluoroethanone ≥97%

Supplier: AOBChem USA

Warning

67898-500MG 67898-1G 67898-5G
77333-054EA 347.11 USD
77333-054 77357-262 77381-180
1-(2,5-Difluoro-4-methoxyphenyl)-2,2-difluoroethanone ≥97%
1-(2,5-Difluoro-4-methoxyphenyl)-2,2-difluoroethanone

Precaución: for Research Use Only

Formula: C₉H₆F₄O₂
MW: 222.136 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28106180
CAS Number: 1823321-97-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR