1-(2-Chloro-4-fluorophenyl)-2,2-difluoroethanone ≥95%

Supplier: AOBChem USA

Warning

67897-500MG 67897-1G 67897-5G 67897-25G
77333-052EA 184.37 USD
77333-052 77347-312 77369-078 77390-190
1-(2-Chloro-4-fluorophenyl)-2,2-difluoroethanone ≥95%
1-(2-Chloro-4-fluorophenyl)-2,2-difluoroethanone

Precaución: for Research Use Only

Formula: C₈H₄ClF₃O
MW: 208.565 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28104776
CAS Number: 2173086-69-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR