2,2-Difluoro-1-(2-(trifluoromethyl)phenyl)ethanone ≥97%
Supplier: AOBChem USA
|
Warning
|
67895-500MG
67895-1G
67895-5G
67895-25G
77333-050EA
136.99
USD
77333-050
77344-628
77369-076
77390-188
2,2-Difluoro-1-(2-(trifluoromethyl)phenyl)ethanone ≥97%
2,2-Difluoro-1-(2-(trifluoromethyl)phenyl)ethanone
Precaución: for Research Use Only
Formula:
C₉H₅F₅O Storage Temperature: Ambient |
MDL Number:
MFCD26045123 CAS Number: 1565499-28-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...