1-(3,4-Difluoro-2-methylphenyl)-2,2-difluoroethanone ≥95%

Supplier: AOBChem USA

Warning

67894-500MG 67894-1G 67894-5G 67894-25G
77333-048EA 157.59 USD
77333-048 77345-664 77369-074 77390-186
1-(3,4-Difluoro-2-methylphenyl)-2,2-difluoroethanone ≥95%
1-(3,4-Difluoro-2-methylphenyl)-2,2-difluoroethanone

Precaución: for Research Use Only

Formula: C₉H₆F₄O
MW: 206.137 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34763056
CAS Number: 2807444-73-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR