2-(Benzyloxy)-3-fluoro-5-methylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

67889-500MG 67889-1G 67889-25G 67889-5G
77333-046EA 285.31 USD
77333-046 77357-258 77390-184 77381-176
2-(Benzyloxy)-3-fluoro-5-methylbenzoic acid ≥97%
2-(Benzyloxy)-3-fluoro-5-methylbenzoic acid

Precaución: for Research Use Only

Formula: C₁₅H₁₃FO₃
MW: 260.26 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34763054
CAS Number: 2807446-98-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR